Anti-BID Rabbit monoclonal antibody unconjugated [clone: EPR9]

Lieferant: Abcam
ABCAAB108293-100EA 785 EUR
ABCAAB108293-100
Anti-BID Rabbit monoclonal antibody unconjugated [clone: EPR9]
Antikörper
Type: Primary
Antigen: BID (BH3 interacting domain death agonist)
Clonality: monoclonal
Clone: EPR9
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR