Anti-ENTPD1 Rabbit monoclonal antibody unconjugated [clone: EPR3678]

Lieferant: Abcam
ABCAAB108248-100EA 920 EUR
ABCAAB108248-100
Anti-ENTPD1 Rabbit monoclonal antibody unconjugated [clone: EPR3678]
Antikörper
Type: Primary
Antigen: ENTPD1 (ectonucleoside triphosphate diphosphohydrolase 1)
Clonality: monoclonal
Clone: EPR3678
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR