Anti-SMYD2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB108217-100EA 885 EUR
ABCAAB108217-100
Anti-SMYD2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SMYD2 (SET and MYND domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR