Anti-ACTN4 Rabbit Monoclonal Antibody [clone: EPR2533]

Lieferant: Abcam
ABCAAB108198-100EA 954 EUR
ABCAAB108198-100
Anti-ACTN4 Rabbit Monoclonal Antibody [clone: EPR2533]
Antikörper
Type: Primary
Antigen: ACTN4 (actinin, alpha 4)
Clonality: monoclonal
Clone: EPR2533
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR