Anti-RYBP Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB107896-100EA 825 EUR
ABCAAB107896-100
Anti-RYBP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RYBP (RING1 and YY1 binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human