Anti-GSK3B Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB107166-100EA 854 EUR
ABCAAB107166-100
Anti-GSK3B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GSK3B (glycogen synthase kinase 3 beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR