Anti-CDCP1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB107165-100EA 798 EUR
ABCAAB107165-100
Anti-CDCP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDCP1 (CUB domain containing protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR