Anti-H3F3AP6 Rat monoclonal antibody [clone: HUGO291]
Lieferant: Abcam
ABCAAB107099-100EA 794 EUR
ABCAAB107099-100
Anti-H3F3AP6 Rat monoclonal antibody [clone: HUGO291]
Type: Primary
Antigen: H3F3AP6 (H3 histone, family 3A, pseudogene 6)
Clonality: monoclonal
Clone: HUGO291
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse