Anti-MORF4L1 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB106818-100EA 835 EUR
ABCAAB106818-100
Anti-MORF4L1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MORF4L1 (mortality factor 4 like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR