Anti-LYRM1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB106665-100EA 797 EUR
ABCAAB106665-100
Anti-LYRM1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LYRM1 (LYR motif containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR