Anti-APC2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB106600-100EA 859 EUR
ABCAAB106600-100
Anti-APC2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: APC2 (adenomatosis polyposis coli 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR