Anti-Endostatin Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: KT60]
Lieferant: Abcam
ABCAAB106491-100EA 840 EUR
ABCAAB106491-100
Anti-Endostatin Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: KT60]
Antikörper
Type: Primary
Antigen: Endostatin
Clonality: monoclonal
Clone: KT60
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human