Anti-LRRTM1 Rabbit polyclonal antibody

Lieferant: Abcam
ABCAAB106414-200EA 0 EUR
ABCAAB106414-200
Anti-LRRTM1 Rabbit polyclonal antibody
Type: Primary
Antigen: LRRTM1 (leucine rich repeat transmembrane neuronal 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR