Anti-ZBED3 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB106383-100EA 837 EUR
ABCAAB106383-100
Anti-ZBED3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZBED3 (zinc finger, BED-type containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR