Anti-Factor alpha XIIa Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB106152-150EA 853 EUR
ABCAAB106152-150
Anti-Factor alpha XIIa Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Factor alpha XIIa
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR