Anti-C3 / C3b Mouse Monoclonal Antibody [clone: 2B10B9B2]

Lieferant: Abcam
ABCAAB106108-50EA 853 EUR
ABCAAB106108-50
Anti-C3 / C3b Mouse Monoclonal Antibody [clone: 2B10B9B2]
Antikörper
Type: Primary
Antigen: C3 / C3b
Clonality: monoclonal
Clone: 2B10B9B2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR