Anti-STAT5A Mouse Monoclonal Antibody [clone: 6.F1]

Lieferant: Abcam
ABCAAB106095-100EA 859 EUR
ABCAAB106095-100
Anti-STAT5A Mouse Monoclonal Antibody [clone: 6.F1]
Antikörper
Type: Primary
Antigen: STAT5A (signal transducer and activator of transcription 5A)
Clonality: monoclonal
Clone: 6.F1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR