Anti-MUC7 Mouse Monoclonal Antibody [clone: 4D2-1D7]

Lieferant: Abcam
ABCAAB105466-50EA 835 EUR
ABCAAB105466-50
Anti-MUC7 Mouse Monoclonal Antibody [clone: 4D2-1D7]
Antikörper
Type: Primary
Antigen: MUC7 (mucin 7, secreted)
Clonality: monoclonal
Clone: 4D2-1D7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR