Anti-TMEM59 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB105417-100EA 797 EUR
ABCAAB105417-100
Anti-TMEM59 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TMEM59 (transmembrane protein 59)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR