Anti-IgG Donkey Polyclonal Antibody (Gold, 18nm)

Lieferant: Abcam
ABCAAB105267-300EA 736 EUR
ABCAAB105267-300
Anti-IgG Donkey Polyclonal Antibody (Gold, 18nm)
Antikörper
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: 18 nm Gold
Epitope:
Host: Donkey
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR