Anti-CLEC9A Rat monoclonal antibody unconjugated [clone: 7H11]

Lieferant: Abcam
ABCAAB104909-50EA 872 EUR
ABCAAB104909-50
Anti-CLEC9A Rat monoclonal antibody unconjugated [clone: 7H11]
Antikörper
Type: Primary
Antigen: CLEC9A (C-type lectin domain family 9, member A)
Clonality: monoclonal
Clone: 7H11
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR