Anti-SIRT1 Mouse Monoclonal Antibody [clone: 1F3]

Lieferant: Abcam
ABCAAB104833-100EA 900 EUR
ABCAAB104833-100
Anti-SIRT1 Mouse Monoclonal Antibody [clone: 1F3]
Antikörper
Type: Primary
Antigen: SIRT1 (sirtuin 1)
Clonality: monoclonal
Clone: 1F3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR