Anti-GABRB3 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB104659-100EA 833 EUR
ABCAAB104659-100
Anti-GABRB3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GABRB3 (gamma-aminobutyric acid (GABA) A receptor, beta 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR