Anti-ITPR2 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB104550-50EA 852 EUR
ABCAAB104550-50
Anti-ITPR2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ITPR2 (inositol 1,4,5-trisphosphate receptor, type 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR