Anti-HBS1L Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB104519-100EA 835 EUR
ABCAAB104519-100
Anti-HBS1L Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: HBS1L (HBS1-like (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR