Anti-PGRMC2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB104437-100EA 835 EUR
ABCAAB104437-100
Anti-PGRMC2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PGRMC2 (progesterone receptor membrane component 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR