Anti-NEUROD2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB104430-100EA 798 EUR
ABCAAB104430-100
Anti-NEUROD2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NEUROD2 (neuronal differentiation 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR