Anti-VR1b Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB104365-100EA 843 EUR
ABCAAB104365-100
Anti-VR1b Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VR1b
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR