Anti-CHEK2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB104232-100EA 882 EUR
ABCAAB104232-100
Anti-CHEK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CHEK2 (checkpoint kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR