Anti-CYB5R1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB104217-50EA 797 EUR
ABCAAB104217-50
Anti-CYB5R1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYB5R1 (cytochrome b5 reductase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR