Anti-LONP1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB103809-100EA 859 EUR
ABCAAB103809-100
Anti-LONP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LONP1 (lon peptidase 1, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR