Anti-Clathrin light chain Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB103553-100EA 852 EUR
ABCAAB103553-100
Anti-Clathrin light chain Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Clathrin light chain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse