Anti-CALCR Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB103422-100EA 840 EUR
ABCAAB103422-100
Anti-CALCR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CALCR (calcitonin receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR