Anti-IgG Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Abcam
ABCAAB102422-500EA 448 EUR
ABCAAB102422-500
Anti-IgG Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Donkey
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR