Anti-IgG/IgM Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB102411-1EA 341 EUR
ABCAAB102411-1
Anti-IgG/IgM Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG+IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR