Anti-IgG Goat Polyclonal Antibody (Biotin)

Lieferant: Abcam
ABCAAB102368-500EA 448 EUR
ABCAAB102368-500
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR