Anti-IgG Goat Polyclonal Antibody (DyLight® 488)

Lieferant: Abcam
ABCAAB102130-500EA 377 EUR
ABCAAB102130-500
Anti-IgG Goat Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Secondary
Antigen: IgG (Pig) (pig IgG)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR