Anti-TRIM62 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB102012-50EA 800 EUR
ABCAAB102012-50
Anti-TRIM62 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRIM62 (tripartite motif containing 62)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR