Anti-PROX1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB101851-100EA 911 EUR
ABCAAB101851-100
Anti-PROX1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PROX1 (prospero homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR