Anti-Piccolo Mouse Monoclonal Antibody [clone: 6H9-B6]

Lieferant: Abcam
ABCAAB101654-100EA 840 EUR
ABCAAB101654-100
Anti-Piccolo Mouse Monoclonal Antibody [clone: 6H9-B6]
Antikörper
Type: Primary
Antigen: Piccolo
Clonality: monoclonal
Clone: 6H9-B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR