Anti-ERBB3 Rabbit monoclonal antibody unconjugated [clone: SP71]

Lieferant: Abcam
ABCAAB101544-7EA 739 EUR
ABCAAB101544-7
Anti-ERBB3 Rabbit monoclonal antibody unconjugated [clone: SP71]
Type: Primary
Antigen: ERBB3 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3)
Clonality: monoclonal
Clone: SP71
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR