Anti-LIFR Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB101228-100EA 882 EUR
ABCAAB101228-100
Anti-LIFR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LIFR (leukemia inhibitory factor receptor alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR