Anti-Melanoma Associated Antigen KBA.62 Mouse monoclonal antibody [clone: KBA.62]
Lieferant: Abcam
ABCAAB101138-250EA 635 EUR
ABCAAB101138-250
Anti-Melanoma Associated Antigen KBA.62 Mouse monoclonal antibody [clone: KBA.62]
Type: Primary
Antigen: Melanoma Associated Antigen KBA.62
Clonality: monoclonal
Clone: KBA.62
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human