Anti-BICC1 Goat polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB101031-100EA 530 EUR
ABCAAB101031-100
Anti-BICC1 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: BICC1 (BicC family RNA binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR