Anti-IRX5 Rabbit polyclonal antibody

Lieferant: Abcam
ABCAAB100939-200EA 0 EUR
ABCAAB100939-200
Anti-IRX5 Rabbit polyclonal antibody
Type: Primary
Antigen: IRX5 (iroquois homeobox 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR