Anti-ABLIM2 Rabbit polyclonal antibody

Lieferant: Abcam
ABCAAB100926-100EA 739 EUR
ABCAAB100926-100
Anti-ABLIM2 Rabbit polyclonal antibody
Type: Primary
Antigen: ABLIM2 (actin binding LIM protein family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR