Anti-GIT2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB100809-100EA 797 EUR
ABCAAB100809-100
Anti-GIT2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GIT2 (G protein-coupled receptor kinase interacting ArfGAP 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR