Anti-EMR1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB100790-100EA 874 EUR
ABCAAB100790-100
Anti-EMR1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EMR1 (egf-like module containing, mucin-like, hormone receptor-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR