Anti-CD19 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB99965-100EA 756 EUR
ABCAAB99965-100
Anti-CD19 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR