Anti-FOXP3 Rabbit monoclonal antibody [clone: SP97]

Lieferant: Abcam
ABCAAB99963-100EA 840 EUR
ABCAAB99963-100
Anti-FOXP3 Rabbit monoclonal antibody [clone: SP97]
Type: Primary
Antigen: FOXP3 (forkhead box P3)
Clonality: monoclonal
Clone: SP97
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR